CO1H324A6522 CombiPhos Catalysts Inc

N-Cyclopropyl-2-(iso-propyl)imidazole-4-boronic acid neopentylglycol ester

  • Cas Number : 2223044-10-6
  • Molecular Weight : 262.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart