CO1H324A652B CombiPhos Catalysts Inc

1-(N-Boc-Pyrrolidin-3-yl)imidazole-4-boronic acid neopentylglycol ester

  • Cas Number : 2223003-65-2
  • Molecular Weight : 349.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart