CO1H324A652C CombiPhos Catalysts Inc

N-Boc-Imidazole-3-boronic acid neopentylglycol ester

  • Cas Number : 2223050-50-6
  • Molecular Weight : 280.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart