CO1H324A6530 CombiPhos Catalysts Inc

N-(Oxan-4-yl)imidazole-3-boronic acid neopentylglycol ester

  • Cas Number : 2223044-17-3
  • Molecular Weight : 264.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart