CO1H324A6535 CombiPhos Catalysts Inc

1-(N-Boc-Pyrrolidin-3-yl)imidazole-3-boronic acid neopentylglycol ester

  • Cas Number : 2223006-26-4
  • Molecular Weight : 349.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart