CO1H324A6542 CombiPhos Catalysts Inc

6-(Oxolan-2-yl)pyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223006-70-8
  • Molecular Weight : 261.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart