CO1H324A6546 CombiPhos Catalysts Inc

5-Trifluoromethylpyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223011-26-3
  • Molecular Weight : 259.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart