CO1H324A6548 CombiPhos Catalysts Inc

5-(Methoxycarbonyl)pyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223040-80-8
  • Molecular Weight : 249.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart