CO1H324A6551 CombiPhos Catalysts Inc

5-(N-Boc-Pyrrolidin-3-yl)pyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223007-09-6
  • Molecular Weight : 360.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart