CO1H324A6552 CombiPhos Catalysts Inc

4-Fluoropyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223033-30-3
  • Molecular Weight : 209.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart