CO1H324A6555 CombiPhos Catalysts Inc

4-(Methoxycarbonyl)pyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223040-85-3
  • Molecular Weight : 249.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart