CO1H324A6557 CombiPhos Catalysts Inc

4-(N-Boc-Piperidin-4-yl)pyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223040-34-2
  • Molecular Weight : 374.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart