CO1H324A655B CombiPhos Catalysts Inc

4-(Oxan-3-yl)pyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2222995-83-5
  • Molecular Weight : 275.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart