CO1H324A655C CombiPhos Catalysts Inc

4-(Oxan-2-yl)pyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223003-57-2
  • Molecular Weight : 275.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart