CO1H324A655E CombiPhos Catalysts Inc

4-(Oxolan-2-yl)pyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223008-65-7
  • Molecular Weight : 261.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart