CO1H324A655F CombiPhos Catalysts Inc

4-(N-Boc-pyrrolidin-3-yl)pyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223009-36-5
  • Molecular Weight : 360.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart