CO1H324A6560 CombiPhos Catalysts Inc

4-Cyclopropylpyridine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223046-60-2
  • Molecular Weight : 231.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart