CO1H324A6563 CombiPhos Catalysts Inc

2-Cyclopropylthiazole-4-boronic acid neopentylglycol ester

  • Cas Number : 2223051-44-1
  • Molecular Weight : 237.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart