CO1H324A6564 CombiPhos Catalysts Inc

2-Cyanothiazole-4-boronic acid neopentylglycol ester

  • Cas Number : 2223040-88-6
  • Molecular Weight : 222.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart