CO1H324A656F CombiPhos Catalysts Inc

6-Cyclopropylpyrazine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223011-39-8
  • Molecular Weight : 232.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart