CO1H324A6570 CombiPhos Catalysts Inc

4-(Cyclohexyl)thiazole-5-boronic acid pinacol ester

  • Cas Number : 2223050-88-0
  • Molecular Weight : 293.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart