CO1H324A6571 CombiPhos Catalysts Inc

6-Cyanopyrazine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223030-94-0
  • Molecular Weight : 217.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart