CO1H324A6576 CombiPhos Catalysts Inc

6-(Oxan-3-yl)pyrazine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223003-58-3
  • Molecular Weight : 276.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart