CO1H324A6583 CombiPhos Catalysts Inc

5-(Oxolan-2-yl)pyrazine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223006-36-6
  • Molecular Weight : 262.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart