CO1H324A6586 CombiPhos Catalysts Inc

5-Bromothiazole-4-boronic acid pinacol ester

  • Cas Number : 2223034-90-8
  • Molecular Weight : 289.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart