CO1H324A658C CombiPhos Catalysts Inc

4-(prop-1-en-2-yl)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 2223011-70-7
  • Molecular Weight : 245.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart