CO1H324A658F CombiPhos Catalysts Inc

2-(prop-1-en-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

  • Cas Number : 2223051-48-5
  • Molecular Weight : 246.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart