CO1H324A6590 CombiPhos Catalysts Inc

methyl 2-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate

  • Cas Number : 1622216-97-0
  • Molecular Weight : 297.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart