CO1H324A6598 CombiPhos Catalysts Inc

6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazine-2-carbonitrile

  • Cas Number : 1186115-53-6
  • Molecular Weight : 231.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart