CO1H324A65B1 CombiPhos Catalysts Inc

2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazine

  • Cas Number : 1310404-63-7
  • Molecular Weight : 236.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart