CO1H324A65BB CombiPhos Catalysts Inc

2-(piperazin-1-yl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazine

  • Cas Number : 1310405-13-0
  • Molecular Weight : 290.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart