CO1H324A65CA CombiPhos Catalysts Inc

4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole-2-carbonitrile

  • Cas Number : 1310384-97-4
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart