CO1H324A65D3 CombiPhos Catalysts Inc

2-(tert-butoxy)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 1310404-55-7
  • Molecular Weight : 277.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart