CO1H324A65E0 CombiPhos Catalysts Inc

4-(6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazin-2-yl)morpholine

  • Cas Number : 1186041-99-5
  • Molecular Weight : 291.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart