CO1H324A65EB CombiPhos Catalysts Inc

6-(Cyclopropylamino)pyridine-2-boronic acid pinacol ester

  • Cas Number : 1309981-36-9
  • Molecular Weight : 260.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart