CO1H324A65EF CombiPhos Catalysts Inc

2-(Cyclopropylamino)thiazole-4-boronic acid pinacol ester

  • Cas Number : 1309981-19-8
  • Molecular Weight : 266.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart