CO1H324A65F0 CombiPhos Catalysts Inc

2-(Cyclobutylamino)thiazole-4-boronic acid pinacol ester

  • Cas Number : 1309982-52-2
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart