CO1H324A65F5 CombiPhos Catalysts Inc

PYRAZINE-2-BORONIC ACID PINACOL ESTER

  • Cas Number : 1083179-99-0
  • Molecular Weight : 206.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart