CO1H324A65FE CombiPhos Catalysts Inc

2-(2-methylpiperidin-1-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole

  • Cas Number : 1310383-64-2
  • Molecular Weight : 308.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart