CO1H324A6602 CombiPhos Catalysts Inc

methyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate

  • Cas Number : 1264176-33-1
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart