CO1H324A661B CombiPhos Catalysts Inc

2-Ethyl-6-methylpyridine-4-boronic acid pinacol ester

  • Cas Number : 2223043-25-0
  • Molecular Weight : 247.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart