CO1H324A661E CombiPhos Catalysts Inc

2-(tert-butyl)-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 2223044-62-8
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart