CO1H324A662A CombiPhos Catalysts Inc

2-Hydroxy-6-trifluoromethylpyridine-4-boronic acid pinacol ester

  • Cas Number : 2223051-19-0
  • Molecular Weight : 289.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart