CO1H324A662D CombiPhos Catalysts Inc

2-Iodo-6-cyanopyridine-4-boronic acid pinacol ester

  • Cas Number : 2223048-26-6
  • Molecular Weight : 355.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart