CO1H324A6630 CombiPhos Catalysts Inc

6-isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile

  • Cas Number : 2223044-21-9
  • Molecular Weight : 272.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart