CO1H324A6632 CombiPhos Catalysts Inc

6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile

  • Cas Number : 2223043-30-7
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart