CO1H324A6634 CombiPhos Catalysts Inc

2-Bromo-6-(methoxycarbonyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223011-87-6
  • Molecular Weight : 342

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart