CO1H324A6638 CombiPhos Catalysts Inc

methyl 6-isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate

  • Cas Number : 2223034-93-1
  • Molecular Weight : 305.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart