CO1H324A663D CombiPhos Catalysts Inc

2-Bromo-6-(sec-butyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223003-00-5
  • Molecular Weight : 340.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart