CO1H324A6643 CombiPhos Catalysts Inc

2-Bromo-6-(2-methoxyphenyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223040-42-2
  • Molecular Weight : 390.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart